missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 100 Well Hinged Tube Storage Box

100 polypropylene storage box designed to hold microcentrifuge tubes and cryogenic vials securely.

Supplier:  Fisherbrand™ 120043F2

 View more versions of this product

Catalog No. 14127156



Only null left

Description

Description

  • The 100-Well Microtube Storage box provides secure storage for microtubes and cryogenic vials
  • Holds up to 100 tubes/vials
  • Quick visual identification with uniform wells and alphanumeric location identification
  • Secure hinged lid with fitted snap clasp
  • Includes a frosted writing area for labeling
  • Made of translucent, vibrantly colored polypropylene
  • Tube capacity is marked on the outside of the box
  • Stackable and lightweight
  • Can be used in temperatures down to -80°C
  • Available in a variety of different colors
Specifications

Chemical Identifiers

Hinged Lid
Square
100 Tubes and Vials
Non-sterile
-80°C
Polypropylene
Cryo Storage Box
5 Pk.
10 x 10
5.6 x 6 x 2.2 in.

Specifications

Hinged Lid
Polypropylene
Cryo Storage Box
5 Pk.
10 x 10
5.6 x 6 x 2.2 in.
Black
Square
100 Tubes and Vials
Non-sterile
-80°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions