Learn More
Fisherbrand™ 100 Well Hinged Tube Storage Box
Catalog No. 14127125
Change view
Click to view available options
Color:
Assorted (Blue, Green, Orange, Purple, Yellow)
Black
Natural
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
14127125 Assorted (Blue, Green, Orange, Purple, Yellow)
14127156 Black
14127183 Natural
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14127125 Supplier Fisherbrand™ Supplier No. 120044F2
Only null left
100 polypropylene storage box designed to hold microcentrifuge tubes and cryogenic vials securely.
  • The 100-Well Microtube Storage box provides secure storage for microtubes and cryogenic vials
  • Holds up to 100 tubes/vials
  • Quick visual identification with uniform wells and alphanumeric location identification
  • Secure hinged lid with fitted snap clasp
  • Includes a frosted writing area for labeling
  • Made of translucent, vibrantly colored polypropylene
  • Tube capacity is marked on the outside of the box
  • Stackable and lightweight
  • Can be used in temperatures down to -80°C
  • Available in a variety of different colors
Specifications
Closure Type Hinged Lid
Color Assorted (Blue, Green, Orange, Purple, Yellow)
Material Polypropylene
Shape Square
Type Cryo Storage Box
Holds 100 Tubes and Vials
Quantity 5 Pk.
Sterility Non-sterile
Tube Array 10 x 10
Withstand Temperature (Metric) -80°C
Dimensions (L x W x H) 5.6 x 6 x 2.2 in.
Show More Show Less