missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 100% Graphite Ferrules

Catalog No. 03050211
Change view
Click to view available options
For Use With (Equipment):
For Varian, Perkin-Elmer
Description:
0.25 in. O.D.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Description
03050211 For Varian, Perkin-Elmer 0.25 in. O.D.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03050211 Supplier Thermo Scientific™ Supplier No. 29033405

Composed of a soft material that is porous to oxygen, making them suitable for most applications except GC/MS interface connections

  • Easy-to-use
  • Form a soft grip with the column and provide a stable seal

Specifications

For Use With (Equipment) For Varian, Perkin-Elmer
Description 0.25 in. O.D.