Learn More
National Safety Apparel™ 100 Cal ArcGuard™ Nomex/Kevlar Bib Overalls
Provides lightweight, durable, heat and flame-resistant solutions. Made of 34% aramid, 33% lyocell, 31% modacrylic, and 2% anti-static materials, Nomex™ is engineered to absorb moisture and dry quickly
$2611.04 - $3475.28
Specifications
Closure Type Buckle
Color Brown
Material Multi-layer DuPont™ Nomex™, Kevlar™
Seam Type Stitched
Suspender Adjustable
View More Specs
Products 8
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
17001301
View Documents
National Safety Apparel™
C45KDQESM32
Small
Each for $2,611.04
Only null left
17001302
View Documents
National Safety Apparel™
C45KDQEMD32
Medium
Each for $2,611.04
Only null left
17001303
View Documents
National Safety Apparel™
C45KDQELG32
Large
Each for $2,611.04
Only null left
17001304
View Documents
National Safety Apparel™
C45KDQEXL32
X-Large
Each for $2,611.04
Only null left
17001305
View Documents
National Safety Apparel™
C45KDQE2X32
2X-Large
Each for $2,872.15
Only null left
17001306
View Documents
National Safety Apparel™
C45KDQE3X32
3X-Large
Each for $2,872.15
Only null left
17001307
View Documents
National Safety Apparel™
C45KDQE4X32
4X-Large
Each for $3,159.35
Only null left
17001308
View Documents
National Safety Apparel™
C45KDQE5X32
5X-Large
Each for $3,475.28
Only null left
Description
Description
  • Arc Rating: 100 cal/cm²
  • CAT 4
  • Fabric/Material: Multilayer DuPont™ Nomex™ and Kevlar™
  • FR fabric will not melt, drip, or ignite
  • FR hook and loop calf closure for easy boot entry
  • Quick disconnect and easy adjustable shoulder straps for quick entry Inherently flame resistant for FR protection
  • Side slant pockets on bib overalls
  • Expandable leg opening to fit over boots
  • Garments embroidered with arc rating Available in a convenient kit
  • Made in the USA
Specifications
Specifications
Buckle
Multi-layer DuPont™ Nomex™, Kevlar™
Adjustable
100 cal/cm²
Flame
Reusable
Brown
Stitched
Open
Standard
CAT 4, NFPA 70 E, CSA Z462, ASTM F1506, OSHA 1910.269
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'