Learn More
National Safety Apparel™ 100 Cal ArcGuard™ Nomex/Kevlar Arc Flash Coat
Kelvar coat complies with OSHA 1910.269; NFPA 70E-2015 Complaint for Arc Flash
Supplier: National Safety Apparel™ C04KDQE032X32
Description
- FR Fabric will NOT melt, drip, or ignite
- FR Hook and loop front closure
- Inherently Flame Resistant
- FR Knit wrist cuffs
- Generous cut allows for unrestrictive movement
Specifications
| FR Hook and Loop | |
| Reusable | |
| DuPont™ Nomex™, Kevlar™ | |
| 2X-Large | |
| Stitched | |
| CAT 4, NFPA 70 E, CSA Z462, ASTM F1506, OSHA 1910.269 |
| Brown | |
| Unisex | |
| Collared | |
| Flame | |
| 100 cal/cm² |