missing translation for 'onlineSavingsMsg'
Learn More

10-Undecyn-1-ol 95.0+%, TCI America™

Catalog No. U00555G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U00555G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. U00555G Supplier TCI America Supplier No. U00555G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2774-84-7
Molecular Formula C11H20O
Molecular Weight (g/mol) 168.28
MDL Number MFCD00041675
InChI Key YUQZOUNRPZBQJK-UHFFFAOYSA-N
Synonym 10-undecyn-1-ol, 10-undecynol, 11-hydroxyundecyne, 1-undecyn-11-ol, acmc-1cmhp, 11-hydroxy-1-undecyne, 10-undecyn-1-ol,, 10-undecyn-1-ol gc
PubChem CID 76015
IUPAC Name undec-10-yn-1-ol
SMILES C#CCCCCCCCCCO

Specifications

Color Yellow
Boiling Point 112°C
Quantity 5 g
Formula Weight 168.28
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 10-Undecyn-1-ol