missing translation for 'onlineSavingsMsg'
Learn More

10-Undecyn-1-ol, 96%

Catalog No. p-7045735
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1180703 1 g
AAL1180706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1180703 Supplier Thermo Scientific Chemicals Supplier No. L1180703

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2774-84-7 | C11H20O | 168.28 g/mol

10-Undecyn-1-ol exhibits an antifungal activity. It is also used in the esterification of pentanoic and stearic acids in the presence of lipase. It is also employed as a reagent in the preparation of 11-bromo-undec-1-yne by reacting with carbon tetrabromide and triphenylphosphine. Further, it acts as an intermediate in synthetic chemistry. In addition to this, it serves as a precursor in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
10-Undecyn-1-ol exhibits an antifungal activity. It is also used in the esterification of pentanoic and stearic acids in the presence of lipase. It is also employed as a reagent in the preparation of 11-bromo-undec-1-yne by reacting with carbon tetrabromide and triphenylphosphine. Further, it acts as an intermediate in synthetic chemistry. In addition to this, it serves as a precursor in organic synthesis.

Solubility
Immiscible with water.

Notes
Air sensitive. Incompatible strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 2774-84-7
Molecular Formula C11H20O
Molecular Weight (g/mol) 168.28
MDL Number MFCD00041675
InChI Key YUQZOUNRPZBQJK-UHFFFAOYSA-N
Synonym 10-undecyn-1-ol, 10-undecynol, 11-hydroxyundecyne, 1-undecyn-11-ol, acmc-1cmhp, 11-hydroxy-1-undecyne, 10-undecyn-1-ol,, 10-undecyn-1-ol gc
PubChem CID 76015
IUPAC Name undec-10-yn-1-ol
SMILES C#CCCCCCCCCCO

Specifications

Density 0.873
Boiling Point 110°C to 111°C (4 mmHg)
Refractive Index 1.4565
Quantity 1 g
Beilstein 1702764
Solubility Information Immiscible with water.
Formula Weight 168.28
Percent Purity 96%
Chemical Name or Material 10-Undecyn-1-ol
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only