Learn More
10-Undecenoyl chloride, 98%
CAS: 38460-95-6 | C11H19ClO | 202.72 g/mol
$196.57 - $196.57
Chemical Identifiers
| CAS | 38460-95-6 |
|---|---|
| Molecular Formula | C11H19ClO |
| Molecular Weight (g/mol) | 202.72 |
| MDL Number | MFCD00000772 |
| InChI Key | MZFGYVZYLMNXGL-UHFFFAOYSA-N |
| Synonym | 10-undecenoyl chloride, 10-undecylenoyl chloride, 10-undecenoylchloride, omega-undecylenic acid chloride, undecylenoyl chloride, .omega.-undecylenic acid chloride, acmc-1ckgs, 10-hendecenoyl chloride, 10-undecenoic acid chloride, 4-02-00-01615 beilstein handbook reference |
| PubChem CID | 38042 |
| IUPAC Name | undec-10-enoyl chloride |
| SMILES | ClC(=O)CCCCCCCCC=C |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC171581000
|
Thermo Scientific Chemicals
171581000 |
100 mL | Glass bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 38460-95-6 | |
| 202.72 | |
| MZFGYVZYLMNXGL-UHFFFAOYSA-N | |
| 38042 | |
| ClC(=O)CCCCCCCCC=C |
| C11H19ClO | |
| MFCD00000772 | |
| 10-undecenoyl chloride, 10-undecylenoyl chloride, 10-undecenoylchloride, omega-undecylenic acid chloride, undecylenoyl chloride, .omega.-undecylenic acid chloride, acmc-1ckgs, 10-hendecenoyl chloride, 10-undecenoic acid chloride, 4-02-00-01615 beilstein handbook reference | |
| undec-10-enoyl chloride |
Specifications
| 38460-95-6 | |
| 100.0 | |
| 0.9400g/mL | |
| 93°C | |
| 98% | |
| C11H19ClO | |
| H2C=CH(CH2)8COCl | |
| 100 mL | |
| 0.94 | |
| MZFGYVZYLMNXGL-UHFFFAOYSA-N | |
| undec-10-enoyl chloride | |
| 38042 | |
| 98% | |
| 10-Undecenoyl chloride, 98% |
| 97.5 | |
| Yellow | |
| 120°C to 122°C (10.0 mmHg) | |
| Authentic | |
| Glass bottle | |
| 1.4532 to 1.4552 | |
| MFCD00000772 | |
| 02, 459 | |
| 10-undecenoyl chloride, 10-undecylenoyl chloride, 10-undecenoylchloride, omega-undecylenic acid chloride, undecylenoyl chloride, .omega.-undecylenic acid chloride, acmc-1ckgs, 10-hendecenoyl chloride, 10-undecenoic acid chloride, 4-02-00-01615 beilstein handbook reference | |
| ClC(=O)CCCCCCCCC=C | |
| 202.72 | |
| 202.72 | |
| Liquid |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 253-951-0
RTECSNumber : YQ2991000
TSCA : TSCA
RUO – Research Use Only