missing translation for 'onlineSavingsMsg'
Learn More

10-Undecenoyl chloride, 98%

Catalog No. AC171581000
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC171581000 100 mL Glass bottle
AC171585000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC171581000 Supplier Thermo Scientific Chemicals Supplier No. 171581000

CAS: 38460-95-6 | C11H19ClO | 202.72 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 38460-95-6
Molecular Formula C11H19ClO
Molecular Weight (g/mol) 202.72
MDL Number MFCD00000772
InChI Key MZFGYVZYLMNXGL-UHFFFAOYSA-N
Synonym 10-undecenoyl chloride, 10-undecylenoyl chloride, 10-undecenoylchloride, omega-undecylenic acid chloride, undecylenoyl chloride, .omega.-undecylenic acid chloride, acmc-1ckgs, 10-hendecenoyl chloride, 10-undecenoic acid chloride, 4-02-00-01615 beilstein handbook reference
PubChem CID 38042
IUPAC Name undec-10-enoyl chloride
SMILES ClC(=O)CCCCCCCCC=C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Density 0.9400g/mL
Boiling Point 120°C to 122°C (10.0 mmHg)
Flash Point 93°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula H2C=CH(CH2)8COCl
Refractive Index 1.4532 to 1.4552
Quantity 100 mL
Beilstein 02, 459
Specific Gravity 0.94
Formula Weight 202.72
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 10-Undecenoyl chloride, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 253-951-0
RTECSNumber YQ2991000
TSCA TSCA

RUO – Research Use Only