missing translation for 'onlineSavingsMsg'
Learn More

10-Undecenoic acid, 99%

Catalog No. AAA1912230
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1912230 250 g
AAA191220B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1912230 Supplier Thermo Scientific Chemicals Supplier No. A1912230

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 112-38-9 | C11H20O2 | 184.279 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
10-Undecenoic acid is used for the preparation of active pharmaceutical ingredients, cosmetics, perfumes, antidandruff shampoos and antimicrobial powders. It is used for the treatment of skin problems and antifungal agent. It is involved in the production of nylon-11, methyl undecylenate, undecylenic aldehyde and undecylenic alcohol. Further, it is used as musk in perfumes and aromas.

Solubility
Soluble in alcohol, chloroform, diethyl ether, benzene and fixed or volatile oils. Insoluble in hot water and cold water.

Notes
Incompatible with bases, oxidizing agents and reducing agents.

Chemical Identifiers

CAS 112-38-9
Molecular Formula C11H20O2
Molecular Weight (g/mol) 184.279
MDL Number MFCD00004442
InChI Key FRPZMMHWLSIFAZ-UHFFFAOYSA-N
Synonym 10-undecenoic acid, undecylenic acid, desenex, 10-hendecenoic acid, renselin, declid, sevinon, 10-undecylenic acid, 9-undecylenic acid, undecyl-10-enic acid
PubChem CID 5634
ChEBI CHEBI:35045
IUPAC Name undec-10-enoic acid
SMILES C=CCCCCCCCCC(=O)O

Specifications

Melting Point 22°C to 25°C
Density 0.91
Boiling Point 126°C to 128°C (1 mmHg)
Flash Point 160°C (320°F)
Refractive Index 1.4493
Quantity 250 g
Beilstein 1762631
Merck Index 14,9848
Solubility Information Soluble in alcohol,chloroform,diethyl ether,benzene and fixed or volatile oils. Insoluble in hot water and cold water.
Formula Weight 184.28
Percent Purity 99%
Chemical Name or Material 10-Undecenoic acid
Show More Show Less
Hazard Category H315-H319-H500
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 203-965-8
RTECSNumber YQ2975000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only