missing translation for 'onlineSavingsMsg'
Learn More

10-Undecenoic acid, 99%

CAS: 112-38-9 | C11H20O2 | 184.279 g/mol

Supplier:  Thermo Scientific Chemicals A191220B

Quantity 1000 g

 View more versions of this product

Catalog No. AAA191220B

Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
10-Undecenoic acid is used for the preparation of active pharmaceutical ingredients, cosmetics, perfumes, antidandruff shampoos and antimicrobial powders. It is used for the treatment of skin problems and antifungal agent. It is involved in the production of nylon-11, methyl undecylenate, undecylenic aldehyde and undecylenic alcohol. Further, it is used as musk in perfumes and aromas.

Solubility
Soluble in alcohol, chloroform, diethyl ether, benzene and fixed or volatile oils. Insoluble in hot water and cold water.

Notes
Incompatible with bases, oxidizing agents and reducing agents.
Specifications

Chemical Identifiers

112-38-9
184.279
FRPZMMHWLSIFAZ-UHFFFAOYSA-N
5634
undec-10-enoic acid
C11H20O2
MFCD00004442
10-undecenoic acid, undecylenic acid, desenex, 10-hendecenoic acid, renselin, declid, sevinon, 10-undecylenic acid, 9-undecylenic acid, undecyl-10-enic acid
CHEBI:35045
C=CCCCCCCCCC(=O)O

Specifications

10-Undecenoic acid
22°C to 25°C
126°C to 128°C (1 mmHg)
C11H20O2
1000 g
1762631
10-undecenoic acid, undecylenic acid, desenex, 10-hendecenoic acid, renselin, declid, sevinon, 10-undecylenic acid, 9-undecylenic acid, undecyl-10-enic acid
FRPZMMHWLSIFAZ-UHFFFAOYSA-N
undec-10-enoic acid
5634
184.28
112-38-9
0.91
160°C (320°F)
1.4493
MFCD00004442
14,9848
Soluble in alcohol,chloroform,diethyl ether,benzene and fixed or volatile oils. Insoluble in hot water and cold water.
C=CCCCCCCCCC(=O)O
184.279
CHEBI:35045
99%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319-H500

EINECSNumber : 203-965-8

RTECSNumber : YQ2975000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions