missing translation for 'onlineSavingsMsg'
Learn More

10-Undecenal 95.0+%, TCI America™

Supplier:  TCI America U001125ML

Catalog No. U001125ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

10-Undecenal
Yellow
C11H20O
25 mL
OFHHDSQXFXLTKC-UHFFFAOYSA-N
undec-10-enal
8187
≥95.0% (GC)
112-45-8
103°C
MFCD00007032
10-undecenal, undecylenic aldehyde, undecylenaldehyde, hendecenal, 10-undecen-1-al, 1-undecen-10-al, 10-undecylenealdehyde, 10-hendecenal, 10-undecenyl aldehyde, c-11 aldehyde, undecylenic
C=CCCCCCCCCC=O
168.28
168.28
Liquid

Chemical Identifiers

112-45-8
168.28
OFHHDSQXFXLTKC-UHFFFAOYSA-N
8187
C=CCCCCCCCCC=O
C11H20O
MFCD00007032
10-undecenal, undecylenic aldehyde, undecylenaldehyde, hendecenal, 10-undecen-1-al, 1-undecen-10-al, 10-undecylenealdehyde, 10-hendecenal, 10-undecenyl aldehyde, c-11 aldehyde, undecylenic
undec-10-enal
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-2464

RTECSNumber : YQ2835000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions