missing translation for 'onlineSavingsMsg'
Learn More

10-Undecen-1-ol 98.0+%, TCI America™

Catalog No. p-7137854
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U004725ML 25 mL
U0047500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. U004725ML Supplier TCI America Supplier No. U004725ML
Only null left

Chemical Identifiers

CAS 112-43-6
Molecular Formula C11H22O
Molecular Weight (g/mol) 170.296
MDL Number MFCD00004750
InChI Key GIEMHYCMBGELGY-UHFFFAOYSA-N
Synonym 10-undecen-1-ol, undecylenic alcohol, undecylenyl alcohol, 10-undecenol, 1-undecen-11-ol, 10-undecylen-1-ol, 11-hydroxy-1-undecene, omega-undecenyl alcohol, undecen-1-ol, .omega.-undecenyl alcohol
PubChem CID 8185
IUPAC Name undec-10-en-1-ol
SMILES C=CCCCCCCCCCO

Specifications

Color Colorless
Boiling Point 138°C
Quantity 25 mL
Formula Weight 170.30
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 10-Undecen-1-ol
EINECSNumber (2)-0258
RTECSNumber YQ2985000
TSCA Yes