missing translation for 'onlineSavingsMsg'
Learn More

10-Undecen-1-ol, 99%

Catalog No. aaa1400236
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1400236 500 g
AAA1400214 25 g
AAA1400222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1400236 Supplier Thermo Scientific Chemicals Supplier No. A1400236
Only null left

CAS: 112-43-6 | C11H22O | 170.296 g/mol

10-Undecen-1-ol is used as a flavoring agent. It reacts with tosylated beta-cyclodextrin to prepare beta-cyclodextrin undecenylether. It is used in the synthesis of 10-Undecenylacrylat by reacting with acryloyl and triethylamine. It reacts with carbon monoxide in the presence of dicobalt octacarbonyl and pyridine as a base at pressures greater than 150 bar and temperatures in the range of 100 to 2000C to linear and branched polyesters having relatively high molecular weights (Mn> 10,000 g / mol) in good yields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
10-Undecen-1-ol is used as a flavoring agent. It reacts with tosylated beta-cyclodextrin to prepare beta-cyclodextrin undecenylether. It is used in the synthesis of 10-Undecenylacrylat by reacting with acryloyl and triethylamine. It reacts with carbon monoxide in the presence of dicobalt octacarbonyl and pyridine as a base at pressures greater than 150 bar and temperatures in the range of 100 to 2000C to linear and branched polyesters having relatively high molecular weights (Mn> 10,000 g / mol) in good yields.

Solubility
Miscible with alcohol and ether. Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong acids.

Chemical Identifiers

CAS 112-43-6
Molecular Formula C11H22O
Molecular Weight (g/mol) 170.296
MDL Number MFCD00004750
InChI Key GIEMHYCMBGELGY-UHFFFAOYSA-N
Synonym 10-undecen-1-ol, undecylenic alcohol, undecylenyl alcohol, 10-undecenol, 1-undecen-11-ol, 10-undecylen-1-ol, 11-hydroxy-1-undecene, omega-undecenyl alcohol, undecen-1-ol, .omega.-undecenyl alcohol
PubChem CID 8185
IUPAC Name undec-10-en-1-ol
SMILES C=CCCCCCCCCCO

Specifications

Melting Point -3°C
Density 0.848
Boiling Point 132°C to 133°C (15 mmHg)
Flash Point 93°C (199°F)
Refractive Index 1.45
Quantity 500 g
Beilstein 1753225
Solubility Information Miscible with alcohol and ether. Immiscible with water.
Formula Weight 170.3
Percent Purity 99%
Chemical Name or Material 10-Undecen-1-ol
Show More Show Less
Hazard Category H227
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 203-971-0
RTECSNumber YQ2985000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only