Learn More
Mopec™ #10 Sterile Scalpel Blades
High-quality, sterile blades used for anatomic pathology or autopsy.
Supplier:  Mopec™ AJ170
 View more versions of this product
Catalog No. 22445383
Only null left
Description
Description
  • Individually wrapped
  • Fits #3, #3L and #5 scalpel handles
Specifications
Chemical Identifiers
Scalpel
Single-use
100 Blades/Pk.
# 10
Sterile
No
Specifications
Carbon Steel
# 10
Sterile
No
Scalpel
Single-use
100 Blades/Pk.
Product Suggestions
Product Suggestions