missing translation for 'onlineSavingsMsg'
Learn More

10-(Pentafluorophenoxycarbonyl)decyltrimethoxysilane, 95%

Catalog No. AAH34117MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH34117MD 250 mg
AAH3411703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH34117MD Supplier Thermo Scientific Chemicals Supplier No. H34117MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 944721-52-2 | C20H29F5O5Si | 472.524 g/mol

It finds its application in optimized immobilization of lectins using self-assembled monolayers on polysilicon encoded materials for cell tagging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application in optimized immobilization of lectins using self-assembled monolayers on polysilicon encoded materials for cell tagging.

Notes
Moisture sensitive. Store away from oxidizing agents and water/moisture. Keep the container tightly sealed and place it in a cool, dry and well ventilated place. Store under inert gas.

Chemical Identifiers

CAS 944721-52-2
Molecular Formula C20H29F5O5Si
Molecular Weight (g/mol) 472.524
MDL Number MFCD13181915
InChI Key KSYPNQIBGIOYPZ-UHFFFAOYSA-N
Synonym 11-pentafluorophenylundecanoato trimethoxysilane, 10-pentafluorophenoxycarbonyl decyltrimethoxysilane, 2,3,4,5,6-pentafluorophenyl 11-trimethoxysilyl undecanoate
PubChem CID 23582157
IUPAC Name (2,3,4,5,6-pentafluorophenyl) 11-trimethoxysilylundecanoate
SMILES CO[Si](CCCCCCCCCCC(=O)OC1=C(C(=C(C(=C1F)F)F)F)F)(OC)OC

Specifications

Boiling Point 145°C to 150°C (0.05mmHg)
Quantity 250 mg
Refractive Index 1.4370-1.4410
Sensitivity Moisture sensitive
Formula Weight 472.52
Percent Purity 95%
Chemical Name or Material 10-(Pentafluorophenoxycarbonyl)decyltrimethoxysilane
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only