missing translation for 'onlineSavingsMsg'
Learn More
Learn More
10-Nonadecanone 97.0+%, TCI America™
$221.83 - $745.62
Chemical Identifiers
| CAS | 504-57-4 |
|---|---|
| Molecular Formula | C19H38O |
| Molecular Weight (g/mol) | 282.51 |
| MDL Number | MFCD00009584 |
| InChI Key | YUPOCHDBBHTUBJ-UHFFFAOYSA-N |
| Synonym | Caprinone, Dinonyl Ketone |
| PubChem CID | 10441 |
| IUPAC Name | nonadecan-10-one |
| SMILES | CCCCCCCCCC(=O)CCCCCCCCC |
Chemical Identifiers
| 504-57-4 | |
| 282.51 | |
| YUPOCHDBBHTUBJ-UHFFFAOYSA-N | |
| 10441 | |
| CCCCCCCCCC(=O)CCCCCCCCC |
| C19H38O | |
| MFCD00009584 | |
| Caprinone, Dinonyl Ketone | |
| nonadecan-10-one |
Specifications
| 504-57-4 | |
| White | |
| C19H38O | |
| 5 g | |
| YUPOCHDBBHTUBJ-UHFFFAOYSA-N | |
| nonadecan-10-one | |
| 10441 | |
| ≥97.0% (GC) | |
| 10-Nonadecanone |
| 58°C | |
| 156°C | |
| MFCD00009584 | |
| Caprinone, Dinonyl Ketone | |
| CCCCCCCCCC(=O)CCCCCCCCC | |
| 282.51 | |
| 282.51 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes