Learn More
Epredia™ 10% Neutral Buffered Formalin in Seal-Rite Containers
Catalog No. 22220682
Change view
Click to view available options
Product Type:
Formalin
Specimen Storage Containers
Packaging Type:
Bulk
Carton
Tray
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Packaging Type
22220682 Formalin Bulk
361015672 Formalin Carton
361015686 Formalin Tray
2860339 Specimen Storage Containers Tray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 22220682 Supplier Epredia™ Supplier No. 59201
Only null left
Use these containers for the transportation and short-term storage of patient specimens.
Closure System
  • Patented Epredia™ Bio-Tite™ closure system is dual-threaded, making it virtually impossible to cross-thread and minimizing leakage
Pre-Filled Containers
  • Nonsterile containers marked with a lot code and expiration date
  • Comply with all updated federal and state OSHA requirement
  • Labeled with highly visible, bright yellow labels with space for patient information for instant identification to minimize health risks
Ordering Information
  • Clear polystyrene containers have screw-top lids
  • Opaque polyethylene containers have snap-top lids
  • Dry containers available separately
Specifications
Capacity 20 mL
Packaging Type Bulk
Product Type Formalin
Quantity 192/cs