Learn More
Epredia™ Concentrated Neutral Buffered Formalin
Description
- Concentrate reduces storage space and contact with formaldehyde
- One gallon of concentrate to four gallons of distilled or deionized water ensures consistent results with those of the premixed 10% NBF
Specifications
Specifications
| Product Type | Formalin |
| Quantity | 1 gal. |
| For Use With (Application) | Fixation |