Learn More
Epredia™ 10% Neutral Buffered Formalin
Reduce exposure to toxic fumes with the premixed Epredia™ Neutral Buffered Formalin (10%). AFIP formulated.
Supplier:  Epredia™ 5755
 View more versions of this product
Catalog No. 22110614
Only null left
Description
Description
  • Phosphate buffers are used to stabilize the pH between 6.8 and 7.2
  • Stringent quality control ensures a formalin concentration of 10%
Specifications
Chemical Identifiers
Formalin
Specifications
Formalin
55 gal.
Product Suggestions
Product Suggestions