Learn More
Epredia™ 10% Neutral Buffered Formalin
Catalog No. 22050105
Change view
Click to view available options
Quantity:
5 Liter
1 gal.
2.5 gal.
5 gal.
55 gal.
Product Type:
Formalin
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
22050105 5 gal. Formalin
22050104 1 gal. Formalin
5735 5 Liter Formalin
22110869 2.5 gal. Formalin
22110614 55 gal. Formalin
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 22050105 Supplier Epredia™ Supplier No. 5705
Reduce exposure to toxic fumes with the premixed Epredia™ Neutral Buffered Formalin (10%). AFIP formulated.
  • Phosphate buffers are used to stabilize the pH between 6.8 and 7.2
  • Stringent quality control ensures a formalin concentration of 10%
Specifications
Product Type Formalin
Quantity 5 gal.