missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ 10% Neutral Buffered Formalin

Catalog No. 22050104
Change view
Click to view available options
Quantity:
5 Liter
1 gal.
2.5 gal.
5 gal.
55 gal.
Product Type:
Formalin
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
22050104 1 gal. Formalin
22050105 5 gal. Formalin
5735 5 Liter Formalin
22110869 2.5 gal. Formalin
22110614 55 gal. Formalin
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 22050104 Supplier Epredia™ Supplier No. SH5701
  • ONLINE EXCLUSIVE PRICE
    $103.89
    $60.43 / Case of 4 Each

    Save $43.46

    42.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Reduce exposure to toxic fumes with the premixed Epredia™ Neutral Buffered Formalin (10%). AFIP formulated.

  • Phosphate buffers are used to stabilize the pH between 6.8 and 7.2
  • Stringent quality control ensures a formalin concentration of 10%

Specifications

Product Type Formalin
Quantity 1 gal.