missing translation for 'onlineSavingsMsg'
Learn More

10% NBF Clear Solution, StatLab

Catalog No. 22035366
Change view
Click to view available options
Quantity:
1 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22035366 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22035366 Supplier Statlab Supplier No. 0169

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Harmful if swallowed, in contact with skin, or if inhaled

10% Neutral Buffered Formalin solution with a specialized buffering system.

  • Replacing standard 10% Neutral Buffered Formalin with NBF Clear ensures that your equipment stays clear of clogging precipitants
  • Available in our 1 gallon StatGrip rectangular F-style bottles featuring HistoFlow dispensing technology - designed for easy pouring, improved ergonomics, and smarter storage

Specifications

For Use With (Equipment) All Tissue Processors
Grade Chemical Solution
Product Type Fixative
Quantity 1 gal.
Solution Type Formalin
Form Solution