missing translation for 'onlineSavingsMsg'
Learn More

10-Methylphenothiazine, 98%

CAS: 1207-72-3 | C13H11NS | 213.30 g/mol

$101.67 - $317.75

Chemical Identifiers

CAS 1207-72-3
Molecular Formula C13H11NS
Molecular Weight (g/mol) 213.30
MDL Number MFCD00041836
InChI Key QXBUYALKJGBACG-UHFFFAOYSA-N
Synonym 10-methyl-10h-phenothiazine, n-methylphenothiazine, 10h-phenothiazine, 10-methyl, phenothiazine, 10-methyl, acmc-1bo1o, bidd:gt0308, phenothiazide methyl derivative, 10-methylphenothiazine, 10-methyl-10h-phenothiazine #, phenothiazine, 10-methyl-8ci
PubChem CID 71015
IUPAC Name 10-methylphenothiazine
SMILES CN1C2=CC=CC=C2SC2=CC=CC=C12
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2164806
View Documents
Thermo Scientific Chemicals
B2164806
5 g
Each for $101.67
Only null left
 
AAB2164814
View Documents
Thermo Scientific Chemicals
B2164814
25 g
Each for $317.75
Only null left
 
Description

Description

They are used in pharmaceutical manufacturing especially antipsychotic drugs. Also used as a stabilizer against polymerization of acrylates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
They are used in pharmaceutical manufacturing especially antipsychotic drugs. Also used as a stabilizer against polymerization of acrylates.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Chemical Identifiers

1207-72-3
213.30
QXBUYALKJGBACG-UHFFFAOYSA-N
71015
CN1C2=CC=CC=C2SC2=CC=CC=C12
C13H11NS
MFCD00041836
10-methyl-10h-phenothiazine, n-methylphenothiazine, 10h-phenothiazine, 10-methyl, phenothiazine, 10-methyl, acmc-1bo1o, bidd:gt0308, phenothiazide methyl derivative, 10-methylphenothiazine, 10-methyl-10h-phenothiazine #, phenothiazine, 10-methyl-8ci
10-methylphenothiazine

Specifications

1207-72-3
C13H11NS
5 g
Insoluble in water.
CN1C2=CC=CC=C2SC2=CC=CC=C12
213.30
213.3
10-Methylphenothiazine
100°C to 103°C
MFCD00041836
10-methyl-10h-phenothiazine, n-methylphenothiazine, 10h-phenothiazine, 10-methyl, phenothiazine, 10-methyl, acmc-1bo1o, bidd:gt0308, phenothiazide methyl derivative, 10-methylphenothiazine, 10-methyl-10h-phenothiazine #, phenothiazine, 10-methyl-8ci
QXBUYALKJGBACG-UHFFFAOYSA-N
10-methylphenothiazine
71015
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 214-896-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only