missing translation for 'onlineSavingsMsg'
Learn More

10-Methylphenothiazine, 98%

Catalog No. AAB2164806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2164806 5 g
AAB2164814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2164806 Supplier Thermo Scientific Chemicals Supplier No. B2164806
Only null left

CAS: 1207-72-3 | C13H11NS | 213.30 g/mol

They are used in pharmaceutical manufacturing especially antipsychotic drugs. Also used as a stabilizer against polymerization of acrylates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
They are used in pharmaceutical manufacturing especially antipsychotic drugs. Also used as a stabilizer against polymerization of acrylates.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 1207-72-3
Molecular Formula C13H11NS
Molecular Weight (g/mol) 213.30
MDL Number MFCD00041836
InChI Key QXBUYALKJGBACG-UHFFFAOYSA-N
Synonym 10-methyl-10h-phenothiazine, n-methylphenothiazine, 10h-phenothiazine, 10-methyl, phenothiazine, 10-methyl, acmc-1bo1o, bidd:gt0308, phenothiazide methyl derivative, 10-methylphenothiazine, 10-methyl-10h-phenothiazine #, phenothiazine, 10-methyl-8ci
PubChem CID 71015
IUPAC Name 10-methylphenothiazine
SMILES CN1C2=CC=CC=C2SC2=CC=CC=C12

Specifications

Melting Point 100°C to 103°C
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 213.3
Percent Purity 98%
Chemical Name or Material 10-Methylphenothiazine
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 214-896-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only