missing translation for 'onlineSavingsMsg'
Learn More

10-Methylphenothiazine 98.0+%, TCI America™

Catalog No. M25175G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M25175G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M25175G Supplier TCI America Supplier No. M25175G
Only null left

Chemical Identifiers

CAS 1207-72-3
Molecular Formula C13H11NS
Molecular Weight (g/mol) 213.30
MDL Number MFCD00041836
InChI Key QXBUYALKJGBACG-UHFFFAOYSA-N
Synonym 10-methyl-10h-phenothiazine, n-methylphenothiazine, 10h-phenothiazine, 10-methyl, phenothiazine, 10-methyl, acmc-1bo1o, bidd:gt0308, phenothiazide methyl derivative, 10-methylphenothiazine, 10-methyl-10h-phenothiazine #, phenothiazine, 10-methyl-8ci
PubChem CID 71015
IUPAC Name 10-methyl-10H-phenothiazine
SMILES CN1C2=CC=CC=C2SC2=CC=CC=C12

Specifications

Melting Point 102°C
Color White-Yellow
Quantity 5 g
Formula Weight 213.30
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 10-Methylphenothiazine