missing translation for 'onlineSavingsMsg'
Learn More

10-Methoxy-5H-dibenzo[b,f]azepine 98.0+%, TCI America™

Catalog No. M189525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M189525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M189525G Supplier TCI America Supplier No. M189525G

Chemical Identifiers

CAS 11-7-4698
Molecular Formula C15H13NO
Molecular Weight (g/mol) 223.28
MDL Number MFCD00101079
InChI Key ZKHZWXLOSIGIGZ-UHFFFAOYSA-N
Synonym 10-Methoxy-2,2′C-iminostilbene
PubChem CID 78424
IUPAC Name 5-methoxy-11H-benzo[b][1]benzazepine
SMILES COC1=CC2=CC=CC=C2NC3=CC=CC=C31

Specifications

Melting Point 127°C
Color Yellow
Quantity 25 g
Formula Weight 223.28
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 10-Methoxy-5H-dibenzo[b,f]azepine
TSCA No
Recommended Storage Refrigerator