missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ 10 μL Microvolume Filter Tips

Catalog No. p-4102770
Change view
Click to view available options
For Use With (Equipment):
Universal Fit Micro
Tip Style:
Maxymum Recovery, Universal
Microvolume
Universal, Maxymum Recovery
Universal, Microvolume
Format:
Bulk
Rack
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Tip Style Format
14222784 Universal Fit Micro Universal, Maxymum Recovery Rack
14222783 Universal Fit Micro Universal, Microvolume Bulk
14222785 Universal Fit Micro Universal, Microvolume Rack
14222792 Universal Fit Micro Maxymum Recovery, Universal Rack
14222791 Universal Fit Micro Microvolume Bulk
14222793 Universal Fit Micro Microvolume Rack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 14222784 Supplier Axygen™ Supplier No. TF300LRS
Only null left

Axygen Universal Tips designed to fit a wide variety of single and multi-channel pipettes, compatible with most popular brands of pipettes. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.

  • Technologically advanced filter material allows free passage of air while blocking aerosol contamination.
  • Filter membrane contains no leachable additives.
  • Axygen™ Maxymum Recovery™ surface improves accuracy and ensures maximum sample recovery.
  • RNase-/DNase-free
  • Nonpyrogenic
  • Human gDNA- and PCR-inhibition-free
  • Order Info

    • Non-Filtered bulk: 1000 tips/pack, 240 packs/case, Filtered bulk: 1000 tips/pack, 10 packs/case
    • Racked: 96 tips/rack, 10 racks/pack, 5 packs/case
    • Stack Racks: 96 tips/rack, 5 racks/stack,10 stacks/case

    Specifications

    Volume (Metric) 0.5 to 10 μL
    Filtered Filtered
    Sterility Sterile
    Color Clear
    Certifications/Compliance USP 3127
    Clean Claims DNase, Rnase Free
    For Use With (Application) Allows recovery of costly samples with improved precision and accuracy
    Format Rack
    Tip Style Universal, Maxymum Recovery
    For Use With (Equipment) Universal Fit Micro
    Material Polypropylene
    Show More Show Less