missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ 10 μL Automation Tips for Agilent™/Velocity 11 VPrep™ and Bravo, 384 Tip Format
GREENER_CHOICE

Catalog No. 14222887
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility
14222887 Sterile
14222886 Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14222887 Supplier Axygen™ Supplier No. VT38410ULRS
Get Up to $2,500 in Products with Purchase
View all promotions

For use with Agilent™/Velocity11 automation systems

Tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality.

  • Clear polypropylene
  • Certified RNase-/DNase-free and nonpyrogenic
  • Nonpyrogenic
  • Maxymum Recovery™ technology for ultra-low retention
  • Available in nonsterile or sterile

Order Info

10 Racks/Pack, 5 Packs/Case

Specifications

Volume (Metric) 10 μL
Filtered Non-filtered
Sterility Sterile
Color Clear
Certifications/Compliance ANSI
Clean Claims DNase, Rnase Free
For Use With (Application) Non-pyrogenic, used for stringent specifications under strict process controls
Format Rack
Tip Style Automation, Round
Packaging SBS Rack
Quantity 50 Racks/Cs.
For Use With (Equipment) Agilent/Velocity VPrep 16ST, Agilent/Velocity VPrep 96ST, Agilent/Velocity VPrep 384ST, Agilent Bravo 384ST
Material Polypropylene
Show More Show Less