missing translation for 'onlineSavingsMsg'
Learn More

10-Hydroxybenzo[h]quinoline 98.0+%, TCI America™

Catalog No. p-7130701
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H07381G 1 g
H07385G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H07381G Supplier TCI America Supplier No. H07381G
Only null left

Chemical Identifiers

CAS 33155-90-7
Molecular Formula C13H9NO
Molecular Weight (g/mol) 195.221
MDL Number MFCD00142350
InChI Key ZVFJWYZMQAEBMO-UHFFFAOYSA-N
Synonym 10-hydroxybenzo h quinoline, benzo h quinolin-10-ol, 10-hydroxy-benzo h quinoline, benzo h quinolin-10 1h-one, 4-hydroxy-5-azaphenanthrene, acmc-1cr1k, benzo h quinoline-10 1h-one, kesrrrlhhxxbrw-uhfffaoysa-n
PubChem CID 11816407
IUPAC Name 1H-benzo[h]quinolin-10-one
SMILES C1=CC(=O)C2=C3C(=CC=CN3)C=CC2=C1

Specifications

Melting Point 101°C
Quantity 1 g
Formula Weight 195.22
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 10-Hydroxybenzo[h]quinoline