missing translation for 'onlineSavingsMsg'
Learn More
Learn More
10-Hydroxybenzo[h]quinoline 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 101°C |
| Quantity | 1 g |
| Formula Weight | 195.22 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 10-Hydroxybenzo[h]quinoline |
Safety and Handling
| TSCA | No |