missing translation for 'onlineSavingsMsg'
Learn More

10-Deacetylbaccatin III 98.0+%, TCI America™

Catalog No. p-7128124
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4148100MG 100 mg
D4148500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4148100MG Supplier TCI America Supplier No. D4148100MG

Chemical Identifiers

CAS 32981-86-5
Molecular Formula C29H36O10
Molecular Weight (g/mol) 544.597
MDL Number MFCD00132913
InChI Key YWLXLRUDGLRYDR-ZUKOLHLZSA-N
PubChem CID 129316875
SMILES CC1=C2C(C(=O)C3(C(CC4C(C3C(C(C2(C)C)(CC1O)O)OC(=O)C5=CC=CC=C5)(CO4)OC(=O)C)O)C)O

Specifications

Melting Point 234°C
Color White
Quantity 100 mg
Formula Weight 544.60
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 10-Deacetylbaccatin III
TSCA No
Recommended Storage Refrigerator