missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(-)-10-Camphorsulfonyl Chloride 98.0+%, TCI America™
$75.95 - $307.57
Chemical Identifiers
| CAS | 39262-22-1 |
|---|---|
| Molecular Formula | C10H15ClO3S |
| Molecular Weight (g/mol) | 250.737 |
| MDL Number | MFCD00064155 |
| InChI Key | BGABKEVTHIJBIW-XVKPBYJWSA-N |
| Synonym | 1r,4s-7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonyl chloride, l--10-camphorsulfonyl chloride, l---10-camphorsulfonyl chloride, 10-camphorsulfonyl chloride, 1r---camphor-10-sulfonyl chloride, 1r-camphor-10-sulfonic acid chloride, pubchem8092, ksc914s0b, l-10-camphorsulfonyl chloride |
| PubChem CID | 9816396 |
| IUPAC Name | [(1S,4R)-7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl]methanesulfonyl chloride |
| SMILES | CC1(C2CCC1(C(=O)C2)CS(=O)(=O)Cl)C |
Chemical Identifiers
| 39262-22-1 | |
| 250.737 | |
| BGABKEVTHIJBIW-XVKPBYJWSA-N | |
| 9816396 | |
| CC1(C2CCC1(C(=O)C2)CS(=O)(=O)Cl)C |
| C10H15ClO3S | |
| MFCD00064155 | |
| 1r,4s-7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonyl chloride, l--10-camphorsulfonyl chloride, l---10-camphorsulfonyl chloride, 10-camphorsulfonyl chloride, 1r---camphor-10-sulfonyl chloride, 1r-camphor-10-sulfonic acid chloride, pubchem8092, ksc914s0b, l-10-camphorsulfonyl chloride | |
| [(1S,4R)-7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl]methanesulfonyl chloride |
Specifications
| 39262-22-1 | |
| White | |
| MFCD00064155 | |
| 3261 | |
| BGABKEVTHIJBIW-XVKPBYJWSA-N | |
| [(1S,4R)-7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl]methanesulfonyl chloride | |
| 9816396 | |
| ≥98.0% (T) | |
| (-)-10-Camphorsulfonyl Chloride |
| 67°C | |
| C10H15ClO3S | |
| 5 g | |
| 1r,4s-7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonyl chloride, l--10-camphorsulfonyl chloride, l---10-camphorsulfonyl chloride, 10-camphorsulfonyl chloride, 1r---camphor-10-sulfonyl chloride, 1r-camphor-10-sulfonic acid chloride, pubchem8092, ksc914s0b, l-10-camphorsulfonyl chloride | |
| CC1(C2CCC1(C(=O)C2)CS(=O)(=O)Cl)C | |
| 250.737 | |
| 250.74 | |
| Crystalline Powder |
Safety and Handling
TSCA : No