missing translation for 'onlineSavingsMsg'
Learn More

(+)-10-Camphorsulfonyl Chloride 97.0+%, TCI America™

Catalog No. p-7124733
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C09985G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C09985G Supplier TCI America Supplier No. C09985G

Chemical Identifiers

CAS 21286-54-4
Molecular Formula C10H15ClO3S
Molecular Weight (g/mol) 250.737
MDL Number MFCD00064156
InChI Key BGABKEVTHIJBIW-GMSGAONNSA-N
Synonym d +-10-camphorsulfonyl chloride, 1s,4r-7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonyl chloride, 1s-+-camphor-10-sulfonyl chloride, d-2-oxobornane-10-sulphonyl chloride, 1s-+-10-camphorsulfonyl chloride, d-+-10-camphorsulfonyl chloride, camphor-10-sulfonyl chloride, +-camphor-10-sulfonyl chloride, +-camphor-10-sulfonic acid chloride
PubChem CID 88856
IUPAC Name [(1R,4S)-7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl]methanesulfonyl chloride
SMILES CC1(C2CCC1(C(=O)C2)CS(=O)(=O)Cl)C

Specifications

Melting Point 66°C
Color White-Yellow
Quantity 5 g
UN Number 3261
Formula Weight 250.74
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (+)-10-Camphorsulfonyl Chloride