missing translation for 'onlineSavingsMsg'
Learn More

(-)-10-Camphorsulfonimine 98.0+%, TCI America™

Catalog No. C13935G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C13935G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C13935G Supplier TCI America Supplier No. C13935G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 60886-80-8
Molecular Formula C10H15NO2S
Molecular Weight (g/mol) 213.30
MDL Number MFCD00013315,MFCD00064576
InChI Key ZAHOEBNYVSWBBW-UHFFFAOYNA-N
Synonym 1s,7s-10,10-dimethyl-3-thia-4-azatricyclo 5.2.1.0<1,5> dec-4-ene-3,3-dione, 1s,7s-10,10-dimethyl-3??-thia-4-azatricyclo 5.2.1.0 1 ,? dec-4-ene-3,3-dione
PubChem CID 45925640
IUPAC Name 10,10-dimethyl-3λ⁶-thia-4-azatricyclo[5.2.1.0¹,⁵]dec-4-ene-3,3-dione
SMILES CC1(C)C2CCC11CS(=O)(=O)N=C1C2

Specifications

Melting Point 229°C
Color White
Quantity 5 g
Formula Weight 213.30
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material (-)-10-Camphorsulfonimine