missing translation for 'onlineSavingsMsg'
Learn More

10-Bromodecanoic acid, 95%

CAS: 50530-12-6 | C10H19BrO2 | 251.164 g/mol

$80.08 - $316.85

Chemical Identifiers

CAS 50530-12-6
Molecular Formula C10H19BrO2
Molecular Weight (g/mol) 251.164
MDL Number MFCD00014388
InChI Key PGVRSPIEZYGOAD-UHFFFAOYSA-N
Synonym 10-bromo-decanoic acid, decanoic acid, 10-bromo, 10-bromanyldecanoic acid, 10-bromodecanoicacid, acmc-1aonn, 10-bromodecanoic acid., .omega.-bromodecanoic acid, ghl.pd_mitscher_leg0.481, ksc489q1r, 10-bromodecanoic acid
PubChem CID 142712
IUPAC Name 10-bromodecanoic acid
SMILES C(CCCCC(=O)O)CCCCBr
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6029206
View Documents
Thermo Scientific Chemicals
H6029206
5 g
Each for $80.08
Only null left
 
AAH6029214
View Documents
Thermo Scientific Chemicals
H6029214
25 g
Each for $316.85
Only null left
 
Description

Description

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Partly miscible in water.

Notes
Air Sensitive. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

50530-12-6
251.164
PGVRSPIEZYGOAD-UHFFFAOYSA-N
142712
C(CCCCC(=O)O)CCCCBr
C10H19BrO2
MFCD00014388
10-bromo-decanoic acid, decanoic acid, 10-bromo, 10-bromanyldecanoic acid, 10-bromodecanoicacid, acmc-1aonn, 10-bromodecanoic acid., .omega.-bromodecanoic acid, ghl.pd_mitscher_leg0.481, ksc489q1r, 10-bromodecanoic acid
10-bromodecanoic acid

Specifications

50530-12-6
C10H19BrO2
MFCD00014388
10-bromo-decanoic acid, decanoic acid, 10-bromo, 10-bromanyldecanoic acid, 10-bromodecanoicacid, acmc-1aonn, 10-bromodecanoic acid., .omega.-bromodecanoic acid, ghl.pd_mitscher_leg0.481, ksc489q1r, 10-bromodecanoic acid
PGVRSPIEZYGOAD-UHFFFAOYSA-N
10-bromodecanoic acid
142712
95%
38°C to 41°C
Br(CH2)9CO2H
5 g
Partly miscible in water.
C(CCCCC(=O)O)CCCCBr
251.164
251.16
10-Bromodecanoic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only