missing translation for 'onlineSavingsMsg'
Learn More
Learn More
10-Bromodecanoic acid, 95%
Description
It is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as a pharmaceutical intermediate.
Solubility
Partly miscible in water.
Notes
Air Sensitive. Incompatible with oxidizing agents.
It is used as a pharmaceutical intermediate.
Solubility
Partly miscible in water.
Notes
Air Sensitive. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 38°C to 41°C |
| Linear Formula | Br(CH2)9CO2H |
| Quantity | 5 g |
| Solubility Information | Partly miscible in water. |
| Formula Weight | 251.16 |
| Percent Purity | 95% |
| Chemical Name or Material | 10-Bromodecanoic acid |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only