missing translation for 'onlineSavingsMsg'
Learn More

10-Bromo-7H-benzo[c]carbazole 98.0+%, TCI America™

Catalog No. p-7123813
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B48831G 1 g
B48835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B48831G Supplier TCI America Supplier No. B48831G

Chemical Identifiers

CAS 1698-16-4
Molecular Formula C16H10BrN
Molecular Weight (g/mol) 296.167
MDL Number MFCD22054961
InChI Key YHAHNQXQOZYZLP-UHFFFAOYSA-N
PubChem CID 68016353
IUPAC Name 10-bromo-7H-benzo[c]carbazole
SMILES C1=CC=C2C(=C1)C=CC3=C2C4=C(N3)C=CC(=C4)Br

Specifications

Melting Point 129°C
Quantity 1 g
Formula Weight 296.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 10-Bromo-7H-benzo[c]carbazole