missing translation for 'onlineSavingsMsg'
Learn More

(+)-10,2-Camphorsultam 98.0+%, TCI America™

Catalog No. C13245G
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
Catalog No. Quantity
C13245G 5 g
1 missing translation for 'options'

Catalog No. C13245G

missing translation for 'mfr': TCI America C13245G

Only null left

Specifications

CAS 108448-77-7
Melting Point 182°C
Color White
Molecular Formula C10H17NO2S
Quantity 5 g
MDL Number MFCD00151762,MFCD00066271,MFCD00151762,MFCD00151762
Synonym 6s,7as-8,8-dimethylhexahydro-3h-3a,6-methanobenzo c isothiazole 2,2-dioxide
InChI Key DPJYJNYYDJOJNO-UHFFFAOYNA-N
SMILES CC1(C)C2CCC11CS(=O)(=O)NC1C2
IUPAC Name 10,10-dimethyl-3λ⁶-thia-4-azatricyclo[5.2.1.0¹,⁵]decane-3,3-dione
Molecular Weight (g/mol) 215.31
PubChem CID 132274328
Formula Weight 215.31
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (+)-10,2-Camphorsultam
Show More Show Less