missing translation for 'onlineSavingsMsg'
Learn More

10-[2-(2-Methoxyethoxy)ethyl]-10H-phenothiazine 98.0+%, TCI America™

Catalog No. m306810g
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M306810G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M306810G Supplier TCI America Supplier No. M306810G
Only null left

Chemical Identifiers

CAS 2098786-35-5
Molecular Formula C17H19NO2S
Synonym MEEPT

Specifications

Color Yellow
Quantity 10 g
Formula Weight 301.40
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 10-[2-(2-Methoxyethoxy)ethyl]-10H-phenothiazine