missing translation for 'onlineSavingsMsg'
Learn More

10,12-Tricosadiynoic acid, 96%

CAS: 66990-30-5 | C23H38O2 | 346.555 g/mol

Supplier:  Thermo Scientific Chemicals L1023106

 View more versions of this product

Catalog No. AAL1023106


Only null left
Explore available promotions

Description

Description

It is an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an active pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
Specifications

Specifications

10,12-Tricosadiynoic acid
55°C to 59°C
MFCD00041683
1880414
10,12-tricosadiynoic acid, 10,12-tricosadiynoicacid, acmc-209nx8, 10,12-tricosadiynoic acid gc
DIEDVCMBPCRJFQ-UHFFFAOYSA-N
tricosa-10,12-diynoic acid
538457
96%
66990-30-5
C23H38O2
5 g
Light sensitive
Insoluble in water.
CCCCCCCCCCC#CC#CCCCCCCCCC(=O)O
346.555
346.56

Chemical Identifiers

66990-30-5
346.555
DIEDVCMBPCRJFQ-UHFFFAOYSA-N
538457
CCCCCCCCCCC#CC#CCCCCCCCCC(=O)O
C23H38O2
MFCD00041683
10,12-tricosadiynoic acid, 10,12-tricosadiynoicacid, acmc-209nx8, 10,12-tricosadiynoic acid gc
tricosa-10,12-diynoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only