missing translation for 'onlineSavingsMsg'
Learn More

10,12-Tricosadiynoic acid, 96%

Catalog No. AAL1023103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1023103 1 g
AAL1023106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1023103 Supplier Thermo Scientific Chemicals Supplier No. L1023103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 66990-30-5 | C23H38O2 | 346.555 g/mol

It is an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an active pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 66990-30-5
Molecular Formula C23H38O2
Molecular Weight (g/mol) 346.555
MDL Number MFCD00041683
InChI Key DIEDVCMBPCRJFQ-UHFFFAOYSA-N
Synonym 10,12-tricosadiynoic acid, 10,12-tricosadiynoicacid, acmc-209nx8, 10,12-tricosadiynoic acid gc
PubChem CID 538457
IUPAC Name tricosa-10,12-diynoic acid
SMILES CCCCCCCCCCC#CC#CCCCCCCCCC(=O)O

Specifications

Melting Point 55°C to 59°C
Quantity 1 g
Beilstein 1880414
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 346.56
Percent Purity 96%
Chemical Name or Material 10,12-Tricosadiynoic acid
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only