missing translation for 'onlineSavingsMsg'
Learn More
Learn More
10,12-Pentacosadiynoic Acid 97.0+%, TCI America™
$37.29 - $130.48
Chemical Identifiers
| CAS | 66990-32-7 |
|---|---|
| Molecular Formula | C25H42O2 |
| Molecular Weight (g/mol) | 374.61 |
| MDL Number | MFCD00041684 |
| InChI Key | ZPUDRBWHCWYMQS-UHFFFAOYSA-N |
| Synonym | 10,12-pentacosadiynoic acid, 10,12-pentacosadiynoicacid, acmc-209nx9, 10-12-pentacosadiynoic acid, 10,12-pentacosadiynoic acid hplc |
| PubChem CID | 538433 |
| IUPAC Name | pentacosa-10,12-diynoic acid |
| SMILES | CCCCCCCCCCCCC#CC#CCCCCCCCCC(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
P1030100MG
|
TCI America
P1030100MG |
100 mg |
Each for $37.29
|
|
|||||
|
P10301G
|
TCI America
P10301G |
1 g |
Each for $130.48
|
|
|||||
Chemical Identifiers
| 66990-32-7 | |
| 374.61 | |
| ZPUDRBWHCWYMQS-UHFFFAOYSA-N | |
| 538433 | |
| CCCCCCCCCCCCC#CC#CCCCCCCCCC(O)=O |
| C25H42O2 | |
| MFCD00041684 | |
| 10,12-pentacosadiynoic acid, 10,12-pentacosadiynoicacid, acmc-209nx9, 10-12-pentacosadiynoic acid, 10,12-pentacosadiynoic acid hplc | |
| pentacosa-10,12-diynoic acid |
Specifications
| 66990-32-7 | |
| C25H42O2 | |
| 100 mg | |
| ZPUDRBWHCWYMQS-UHFFFAOYSA-N | |
| pentacosa-10,12-diynoic acid | |
| 538433 | |
| ≥97.0% (GC,T) | |
| 10,12-Pentacosadiynoic Acid |
| 65°C | |
| MFCD00041684 | |
| 10,12-pentacosadiynoic acid, 10,12-pentacosadiynoicacid, acmc-209nx9, 10-12-pentacosadiynoic acid, 10,12-pentacosadiynoic acid hplc | |
| CCCCCCCCCCCCC#CC#CCCCCCCCCC(O)=O | |
| 374.61 | |
| 374.61 | |
| Crystalline Powder |
Safety and Handling
TSCA : No