missing translation for 'onlineSavingsMsg'
Learn More

10,11-Dihydrocarbamazepine, 99%

Catalog No. p-7047019
Change view
Click to view available options
Quantity:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL17271MF 50 mg
AAL17271MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL17271MF Supplier Thermo Scientific Chemicals Supplier No. L17271MF
Only null left

CAS: 3564-73-6 | C15H14N2O | 238.29 g/mol

10,11-Dihydrocarbamazepine is used as internal standard during simultaneous determination of carbamazepine and its active metabolite in plasma by liquid chromatography. It can be used in chemiluminescence method for determination of carbamazepine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
10,11-Dihydrocarbamazepine is used as internal standard during simultaneous determination of carbamazepine and its active metabolite in plasma by liquid chromatography. It can be used in chemiluminescence method for determination of carbamazepine.

Solubility
Soluble in dichloromethane. Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 3564-73-6
Molecular Formula C15H14N2O
Molecular Weight (g/mol) 238.29
MDL Number MFCD00005072
InChI Key PHNLCHMJDSSPDQ-UHFFFAOYSA-N
Synonym 10,11-dihydrocarbamazepine, 10,11-dihydro-5h-dibenzo b,f azepine-5-carboxamide, unii-ec017va9fr, dihydrocarbamazepine, ec017va9fr, 5h-dibenz b,f azepine-5-carboxamide, 10,11-dihydro, chembl41543, 5,6-dihydrobenzo b 1 benzazepine-11-carboxamide, 10,11-dihydro-5h-dibenz b,f azepine-5-carboxamide
PubChem CID 19099
IUPAC Name 2-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaene-2-carboxamide
SMILES NC(=O)N1C2=CC=CC=C2CCC2=CC=CC=C12

Specifications

Melting Point 204°C to 206°C
Quantity 50 mg
Beilstein 196969
Solubility Information Soluble in dichloromethane. Insoluble in water.
Formula Weight 238.29
Percent Purity 99%
Chemical Name or Material 10,11-Dihydrocarbamazepine
EINECSNumber 222-649-0
RTECSNumber HN8300000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only