missing translation for 'onlineSavingsMsg'
Learn More

10,11-Dihydro-5H-dibenzo[b,f]azepine 97.0+%, TCI America™

Catalog No. D026925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D026925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D026925G Supplier TCI America Supplier No. D026925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 494-19-9
Molecular Formula C14H13N
Molecular Weight (g/mol) 195.27
MDL Number MFCD00005070
InChI Key ZSMRRZONCYIFNB-UHFFFAOYSA-N
Synonym iminodibenzyl, 10,11-dihydro-5h-dibenzo b,f azepine, 10,11-dihydro-5h-dibenz b,f azepine, iminobibenzyl, 2,2'-iminodibenzyl, 2,2'-iminobibenzyl, iminodibenyl, 5h-dibenz b,f azepine, 10,11-dihydro, unii-262bx7oe3u, 10,11-dihydro-5-dibenz b,f azepine
PubChem CID 10308
IUPAC Name 2-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaene
SMILES C1CC2=CC=CC=C2NC2=CC=CC=C12

Specifications

Melting Point 107°C
Quantity 25 g
Formula Weight 195.27
Percent Purity ≥97.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 10,11-Dihydro-5H-dibenzo[b,f]azepine
EINECSNumber (5)-3872
RTECSNumber HN8950000
TSCA Yes