missing translation for 'onlineSavingsMsg'
Learn More
Learn More
10,11-Dihydro-5H-dibenzo[b,f]azepine-5-carbonyl Chloride 98.0+%, TCI America™
$61.61 - $188.67
Chemical Identifiers
| CAS | 33948-19-5 |
|---|---|
| Molecular Formula | C15H12ClNO |
| Molecular Weight (g/mol) | 257.717 |
| MDL Number | MFCD00012314 |
| InChI Key | COHHZMJBMIHLGF-UHFFFAOYSA-N |
| PubChem CID | 118579 |
| IUPAC Name | 5,6-dihydrobenzo[b][1]benzazepine-11-carbonyl chloride |
| SMILES | C1CC2=CC=CC=C2N(C3=CC=CC=C31)C(=O)Cl |
Chemical Identifiers
| 33948-19-5 | |
| 257.717 | |
| COHHZMJBMIHLGF-UHFFFAOYSA-N | |
| 5,6-dihydrobenzo[b][1]benzazepine-11-carbonyl chloride |
| C15H12ClNO | |
| MFCD00012314 | |
| 118579 | |
| C1CC2=CC=CC=C2N(C3=CC=CC=C31)C(=O)Cl |
Specifications
| 122°C | |
| White-Yellow | |
| MFCD00012314 | |
| 2,2′C-Iminodibenzyl-5-carbonyl Chloride | |
| C1CC2=CC=CC=C2N(C3=CC=CC=C31)C(=O)Cl | |
| 5 g | |
| 118579 | |
| ≥98.0% (HPLC,T) | |
| 10,11-Dihydro-5H-dibenzo[b,f]azepine-5-carbonyl Chloride |
| 33948-19-5 | |
| C15H12ClNO | |
| 3261 | |
| COHHZMJBMIHLGF-UHFFFAOYSA-N | |
| 5,6-dihydrobenzo[b][1]benzazepine-11-carbonyl chloride | |
| 257.717 | |
| 257.72 | |
| Crystalline Powder |
Safety and Handling
TSCA : No