missing translation for 'onlineSavingsMsg'
Learn More

10,10'-Dimethyl-9,9'-biacridinium Bis(monomethyl Terephthalate) [for Chemiluminescence Research], TCI America™

Catalog No. p-7123480
Change view
Click to view available options
Quantity:
1 g
100 mg
Catalog No. Quantity
B4339100MG 100 mg
B43391G 1 g
2 options

Catalog No. B4339100MG

Supplier: TCI America B4339100MG

Only null left

Chemical Identifiers

CAS 469865-01-8
Molecular Formula C46H36N2O8
Molecular Weight (g/mol) 744.8
InChI Key BPSROIZFFFEAQI-UHFFFAOYSA-L
Synonym MMT
PubChem CID 130406107
IUPAC Name 4-methoxycarbonylbenzoate;10-methyl-9-(10-methylacridin-10-ium-9-yl)acridin-10-ium
SMILES C[N+]1=C2C=CC=CC2=C(C3=CC=CC=C31)C4=C5C=CC=CC5=[N+](C6=CC=CC=C64)C.COC(=O)C1=CC=C(C=C1)C(=O)[O-].COC(=O)C1=CC=C(C=C1)C(=O)[O-]

Specifications

Quantity 100 mg
Formula Weight 744.80
Physical Form Crystalline Powder
Chemical Name or Material 10,10′-Dimethyl-9,9′-biacridinium Bis(monomethyl Terephthalate) [for Chemiluminescence R