missing translation for 'onlineSavingsMsg'
Learn More

10,10'-Dibromo-9,9'-bianthracene 98.0+%, TCI America™

Catalog No. D42541G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D42541G 1 g
D42545G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D42541G Supplier TCI America Supplier No. D42541G

Chemical Identifiers

CAS 121848-75-7
Molecular Formula C28H16Br2
Molecular Weight (g/mol) 512.244
MDL Number MFCD11045035
InChI Key NPNNLGXEAGTSRN-UHFFFAOYSA-N
PubChem CID 15195999
IUPAC Name 9-bromo-10-(10-bromoanthracen-9-yl)anthracene
SMILES C1=CC=C2C(=C1)C(=C3C=CC=CC3=C2Br)C4=C5C=CC=CC5=C(C6=CC=CC=C64)Br

Specifications

Melting Point 342°C
Color Yellow
Quantity 1 g
Formula Weight 512.24
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 10,10′-Dibromo-9,9′-bianthracene