missing translation for 'onlineSavingsMsg'
Learn More
Learn More
10-(1,1':3',1″-Terphenyl-5'-yl)anthracene-9-boronic Acid 97.0+%, TCI America™
(contains varying amounts of Anhydride)
$234.87 - $704.62
Chemical Identifiers
| CAS | 1415334-59-6 |
|---|---|
| Molecular Formula | C32H23BO2 |
| Molecular Weight (g/mol) | 450.344 |
| InChI Key | NQOSAYPSOKFYBW-UHFFFAOYSA-N |
| Synonym | 10-(m-Terphenyl-5′C-yl)anthracene-9-boronic Acid |
| PubChem CID | 127255460 |
| IUPAC Name | [10-(3,5-diphenylphenyl)anthracen-9-yl]boronic acid |
| SMILES | B(C1=C2C=CC=CC2=C(C3=CC=CC=C13)C4=CC(=CC(=C4)C5=CC=CC=C5)C6=CC=CC=C6)(O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
T3280200MG
|
TCI America
T3280200MG |
200 mg |
Each for $234.87
|
|
|||||
|
T32801G
|
TCI America
T32801G |
1 g |
Each for $704.62
|
|
|||||
Chemical Identifiers
| 1415334-59-6 | |
| 450.344 | |
| 10-(m-Terphenyl-5′C-yl)anthracene-9-boronic Acid | |
| [10-(3,5-diphenylphenyl)anthracen-9-yl]boronic acid |
| C32H23BO2 | |
| NQOSAYPSOKFYBW-UHFFFAOYSA-N | |
| 127255460 | |
| B(C1=C2C=CC=CC2=C(C3=CC=CC=C13)C4=CC(=CC(=C4)C5=CC=CC=C5)C6=CC=CC=C6)(O)O |
Specifications
| 1415334-59-6 | |
| C32H23BO2 | |
| 10-(m-Terphenyl-5′C-yl)anthracene-9-boronic Acid | |
| B(C1=C2C=CC=CC2=C(C3=CC=CC=C13)C4=CC(=CC(=C4)C5=CC=CC=C5)C6=CC=CC=C6)(O)O | |
| 450.344 | |
| 450.34 | |
| Crystalline Powder |
| Yellow | |
| 200 mg | |
| NQOSAYPSOKFYBW-UHFFFAOYSA-N | |
| [10-(3,5-diphenylphenyl)anthracen-9-yl]boronic acid | |
| 127255460 | |
| ≥97.0% (HPLC) | |
| 10-(1,1′:3′,1′′-Terphenyl-5′-yl)anthracene-9-boronic Acid (contains va |
Safety and Handling
TSCA : No