missing translation for 'onlineSavingsMsg'
Learn More

10-(1,1':3',1″-Terphenyl-5'-yl)anthracene-9-boronic Acid 97.0+%, TCI America™

Catalog No. T3280200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3280200MG 200 mg
T32801G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T3280200MG Supplier TCI America Supplier No. T3280200MG

(contains varying amounts of Anhydride)

Chemical Identifiers

CAS 1415334-59-6
Molecular Formula C32H23BO2
Molecular Weight (g/mol) 450.344
InChI Key NQOSAYPSOKFYBW-UHFFFAOYSA-N
Synonym 10-(m-Terphenyl-5′C-yl)anthracene-9-boronic Acid
PubChem CID 127255460
IUPAC Name [10-(3,5-diphenylphenyl)anthracen-9-yl]boronic acid
SMILES B(C1=C2C=CC=CC2=C(C3=CC=CC=C13)C4=CC(=CC(=C4)C5=CC=CC=C5)C6=CC=CC=C6)(O)O

Specifications

Color Yellow
Quantity 200 mg
Formula Weight 450.34
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 10-(1,1′:3′,1′′-Terphenyl-5′-yl)anthracene-9-boronic Acid (contains va